Assay Details
Gene Symbol: LOC5567565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657506.2 | XP_001657556.2 | 2 - 3 | 278 | 74 | |
GenBank mRNA | AY432885.1 | - | 2 - 3 | 197 | 74 | |
CR938042.1 | - | 2 - 3 | 197 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
