Assay Details
Gene Symbol: LOC5567565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657506.2 | XP_001657556.2 | 3 - 4 | 399 | 63 | |
GenBank mRNA | AY432885.1 | - | 3 - 4 | 318 | 63 | |
CR938042.1 | - | 3 - 4 | 318 | 63 | ||
M95187.1 | - | 3 - 4 | 307 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
