Assay Details
Gene Symbol: LOC5567565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657506.2 | XP_001657556.2 | 4 - 5 | 524 | 69 | |
GenBank mRNA | AY432885.1 | - | 4 - 5 | 443 | 69 | |
CR938042.1 | - | 4 - 5 | 443 | 69 | ||
M95187.1 | - | 4 - 5 | 432 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
