Assay Details
Gene Symbol: LOC5574421 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661388.2 | XP_001661438.1 | 1 - 2 | 70 | ||
GenBank mRNA | AY432886.1 | - | 1 - 2 | 46 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001661388.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
