Assay Details
Gene Symbol: LOC5574421 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661388.2 | XP_001661438.1 | 2 - 3 | 67 | ||
GenBank mRNA | AY432886.1 | - | 2 - 3 | 175 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
