Assay Details
Gene Symbol: LOC5575813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662189.3 | XP_001662239.2 | 3 - 4 | 414 | 74 | |
XM_021849765.1 | XP_021705457.1 | 3 - 4 | 483 | 74 | ||
XM_021849766.1 | XP_021705458.1 | 3 - 4 | 610 | 74 | ||
GenBank mRNA | AY432890.1 | - | 3 - 4 | 478 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
