Assay Details
Gene Symbol: LOC5563689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001647909.2 | XP_001647959.1 | 1 - 2 | 409 | 61 | |
GenBank mRNA | AY432935.1 | - | 397 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
