Assay Details
Gene Symbol: LOC5563689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001647909.2 | XP_001647959.1 | 2 - 3 | 682 | 114 | |
GenBank mRNA | AY432935.1 | - | 670 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
