Assay Details
Gene Symbol: LOC5563671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001647914.2 | XP_001647964.1 | 3 - 4 | 571 | 60 | |
GenBank mRNA | AY432952.1 | - | 3 - 4 | 552 | 60 | |
CR938158.1 | - | 3 - 4 | 524 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
