Assay Details
Gene Symbol: LOC5576745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662794.2 | XP_001662844.1 | 2 - 3 | 304 | 87 | |
GenBank mRNA | DQ440444.1 | - | 1 - 2 | 109 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
