Assay Details
Gene Symbol: LOC5580248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844522.1 | XP_021700214.1 | 4 - 5 | 82 | ||
XM_021844523.1 | XP_021700215.1 | 5 - 6 | 82 | |||
GenBank mRNA | DQ459411.1 | - | 3 - 4 | 410 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
