Assay Details
Gene Symbol: LOC5578456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656927.3 | XP_001656977.3 | 1 - 2 | 248 | 94 | |
XM_021851331.1 | XP_021707023.1 | 3 - 4 | 1379 | 94 | ||
XM_021851332.1 | XP_021707024.1 | 3 - 4 | 2045 | 94 | ||
XM_021851333.1 | XP_021707025.1 | 3 - 4 | 2040 | 94 | ||
XM_021851334.1 | XP_021707026.1 | 3 - 4 | 1374 | 94 | ||
XM_021851336.1 | XP_021707028.1 | 3 - 4 | 928 | 94 | ||
XM_021851337.1 | XP_021707029.1 | 3 - 4 | 535 | 94 | ||
XM_021851338.1 | XP_021707030.1 | 2 - 3 | 782 | 94 | ||
GenBank mRNA | EF209048.1 | - | 2 - 3 | 699 | 94 |
