Assay Details
Gene Symbol: LOC5578456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656927.3 | XP_001656977.3 | 2 - 3 | 387 | 79 | |
XM_021851331.1 | XP_021707023.1 | 4 - 5 | 1518 | 79 | ||
XM_021851332.1 | XP_021707024.1 | 4 - 5 | 2184 | 79 | ||
XM_021851333.1 | XP_021707025.1 | 4 - 5 | 2179 | 79 | ||
XM_021851334.1 | XP_021707026.1 | 4 - 5 | 1513 | 79 | ||
XM_021851336.1 | XP_021707028.1 | 4 - 5 | 1067 | 79 | ||
XM_021851337.1 | XP_021707029.1 | 4 - 5 | 674 | 79 | ||
XM_021851338.1 | XP_021707030.1 | 3 - 4 | 921 | 79 | ||
XM_021851339.1 | XP_021707031.1 | 4 - 5 | 1495 | 79 | ||
GenBank mRNA | EF209048.1 | - | 3 - 4 | 838 | 79 |
