Assay Details
Gene Symbol: LOC5578456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656927.3 | XP_001656977.3 | 5 - 6 | 1841 | 115 | |
XM_021851331.1 | XP_021707023.1 | 7 - 8 | 2972 | 115 | ||
XM_021851332.1 | XP_021707024.1 | 7 - 8 | 3638 | 115 | ||
XM_021851333.1 | XP_021707025.1 | 7 - 8 | 3633 | 115 | ||
XM_021851334.1 | XP_021707026.1 | 7 - 8 | 2967 | 115 | ||
XM_021851336.1 | XP_021707028.1 | 7 - 8 | 2521 | 115 | ||
XM_021851337.1 | XP_021707029.1 | 7 - 8 | 2128 | 115 | ||
XM_021851338.1 | XP_021707030.1 | 6 - 7 | 2375 | 115 | ||
XM_021851339.1 | XP_021707031.1 | 7 - 8 | 2949 | 115 | ||
GenBank mRNA | EF209048.1 | - | 6 - 7 | 2292 | 115 |

More Information
Set Membership: |
![]() ![]() |
