Assay Details
Gene Symbol: LOC5578673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657021.2 | XP_001657071.2 | 1 - 2 | 1050 | 74 | |
GenBank mRNA | EU267007.1 | - | 1 - 2 | 369 | 74 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5578673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657021.2 | XP_001657071.2 | 1 - 2 | 1050 | 74 | |
GenBank mRNA | EU267007.1 | - | 1 - 2 | 369 | 74 |