Assay Details
Gene Symbol: LOC5578673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657021.2 | XP_001657071.2 | 2 - 3 | 2026 | 70 | |
GenBank mRNA | EU267007.1 | - | 2 - 3 | 1345 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
