Assay Details
Gene Symbol: LOC5578673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657021.2 | XP_001657071.2 | 3 - 4 | 108 | ||
GenBank mRNA | EU267007.1 | - | 3 - 4 | 1616 | 108 |

More Information
Set Membership: |
![]() ![]() |
