Assay Details
Gene Symbol: LOC5578664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851370.1 | XP_021707062.1 | 3 - 4 | 984 | 107 | |
GenBank mRNA | EU267008.1 | - | 3 - 4 | 856 | 107 |

More Information
Set Membership: |
![]() ![]() |
