Assay Details
Gene Symbol: LOC5578664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851370.1 | XP_021707062.1 | 5 - 6 | 1724 | 85 | |
GenBank mRNA | EU267008.1 | - | 5 - 6 | 1596 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
