Assay Details
Gene Symbol: LOC5578666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849548.1 | XP_021705240.1 | 5 - 6 | 1816 | 93 | |
XM_021849549.1 | XP_021705241.1 | 5 - 6 | 1718 | 93 | ||
GenBank mRNA | EU267009.1 | - | 4 - 5 | 1517 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
