Assay Details
Gene Symbol: LOC5578663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855164.1 | XP_021710856.1 | 2 - 3 | 956 | 101 | |
XM_021855165.1 | XP_021710857.1 | 2 - 3 | 912 | 101 | ||
XM_021855166.1 | XP_021710858.1 | 2 - 3 | 893 | 101 | ||
XM_021855167.1 | XP_021710859.1 | 2 - 3 | 849 | 101 | ||
GenBank mRNA | EU267011.1 | - | 1 - 2 | 653 | 101 |

More Information
Set Membership: |
![]() ![]() |
