Assay Details
Gene Symbol: LOC5578663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855164.1 | XP_021710856.1 | 5 - 6 | 1976 | 91 | |
XM_021855165.1 | XP_021710857.1 | 5 - 6 | 1932 | 91 | ||
XM_021855166.1 | XP_021710858.1 | 5 - 6 | 1913 | 91 | ||
XM_021855167.1 | XP_021710859.1 | 5 - 6 | 1869 | 91 | ||
GenBank mRNA | EU267011.1 | - | 4 - 5 | 1673 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
