Assay Details
Gene Symbol: LOC5567562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847260.1 | XP_021702952.1 | 1 - 2 | 1042 | 73 | |
GenBank mRNA | EU604100.1 | - | 1 - 2 | 437 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
