Assay Details
Gene Symbol: LOC5570234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658963.2 | XP_001659013.1 | 4 - 5 | 695 | 80 | |
XM_021855794.1 | XP_021711486.1 | 5 - 6 | 651 | 80 | ||
GenBank mRNA | FJ357570.1 | - | 4 - 5 | 458 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
