Assay Details
Gene Symbol: LOC5566108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839745.1 | XP_021695437.1 | 2 - 3 | 432 | 63 | |
XM_021839746.1 | XP_021695438.1 | 2 - 3 | 431 | 63 | ||
XM_021839747.1 | XP_021695439.1 | 2 - 3 | 455 | 63 | ||
XM_021839748.1 | XP_021695440.1 | 2 - 3 | 430 | 63 | ||
XM_021839749.1 | XP_021695441.1 | 2 - 3 | 428 | 63 | ||
XM_021839750.1 | XP_021695442.1 | 2 - 3 | 427 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
