Assay Details
Gene Symbol: LOC5569281 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843990.1 | XP_021699682.1 | 3 - 4 | 1693 | 67 | |
XM_021843992.1 | XP_021699684.1 | 4 - 5 | 1624 | 67 | ||
XM_021843993.1 | XP_021699685.1 | 4 - 5 | 1555 | 67 | ||
XM_021843994.1 | XP_021699686.1 | 4 - 5 | 1491 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
