Assay Details
Gene Symbol: LOC5576000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849024.1 | XP_021704716.1 | 4 - 5 | 2388 | 72 | |
GenBank mRNA | AF303224.1 | - | 4 - 5 | 2375 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
