Assay Details
Gene Symbol: LOC5574485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655096.2 | XP_001655146.1 | 3 - 4 | 447 | 67 | |
GenBank mRNA | AF315552.1 | - | 3 - 4 | 342 | 67 | |
AY133344.1 | - | 2 - 3 | 335 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
