Assay Details
Gene Symbol: LOC5570613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850734.1 | XP_021706426.1 | 3 - 4 | 565 | 61 | |
XM_021850739.1 | XP_021706431.1 | 3 - 4 | 567 | 61 | ||
GenBank mRNA | AF369383.1 | - | 3 - 4 | 296 | 61 | |
AF543193.1 | - | 3 - 4 | 368 | 61 | ||
AY330296.1 | - | 2 - 3 | 479 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
