Assay Details
Gene Symbol: LOC5570613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850734.1 | XP_021706426.1 | 6 - 7 | 1173 | 110 | |
GenBank mRNA | AF369383.1 | - | 6 - 7 | 904 | 110 | |
AF543193.1 | - | 6 - 7 | 976 | 110 | ||
AY330296.1 | - | 5 - 6 | 1087 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
