Assay Details
Gene Symbol: LOC5565400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649675.3 | XP_001649725.2 | 1 - 2 | 74 | ||
GenBank mRNA | AF396870.1 | - | 1 - 2 | 297 | 74 | |
AF396871.1 | - | 1 - 2 | 74 | |||
AY133346.1 | - | 1 - 2 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
