Assay Details
Gene Symbol: LOC5578243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663715.2 | XP_001663765.1 | 1 - 2 | 156 | 75 | |
GenBank mRNA | AF399675.1 | - | 1 - 2 | 36 | 75 | |
AY064121.1 | - | 1 - 2 | 36 | 75 | ||
AY433188.1 | - | 1 - 2 | 75 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001663715.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
