Assay Details
Gene Symbol: LOC5569427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658467.3 | XP_001658517.2 | 4 - 5 | 1943 | 85 | |
XM_021841875.1 | XP_021697567.1 | 4 - 5 | 1250 | 85 | ||
GenBank mRNA | AF498104.1 | - | 3 - 4 | 832 | 85 | |
AF498105.1 | - | 4 - 5 | 1092 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
