Assay Details
Gene Symbol: LOC5569427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658467.3 | XP_001658517.2 | 5 - 6 | 2474 | 76 | |
XM_021841875.1 | XP_021697567.1 | 5 - 6 | 1781 | 76 | ||
GenBank mRNA | AF498104.1 | - | 4 - 5 | 1363 | 76 | |
AF498105.1 | - | 5 - 6 | 1623 | 76 | ||
AF498106.1 | - | 2 - 3 | 1072 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
