Assay Details
Gene Symbol: LOC5575011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494871.2 | XP_011493173.2 | 2 - 3 | 801 | 93 | |
XM_021852568.1 | XP_021708260.1 | 4 - 5 | 1095 | 93 | ||
XM_021852569.1 | XP_021708261.1 | 4 - 5 | 1232 | 93 | ||
GenBank mRNA | AY326254.1 | - | 4 - 5 | 1200 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
