Assay Details
Gene Symbol: LOC5571503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659703.2 | XP_001659753.1 | 1 - 2 | 301 | 92 | |
GenBank mRNA | AY431109.1 | - | 1 - 2 | 288 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
