Assay Details
Gene Symbol: LOC5571503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659703.2 | XP_001659753.1 | 3 - 4 | 567 | 95 | |
GenBank mRNA | AY431109.1 | - | 3 - 4 | 554 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
