Assay Details
Gene Symbol: LOC5575683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655759.2 | XP_001655809.1 | 1 - 2 | 437 | 83 | |
XM_001655760.2 | XP_001655810.1 | 1 - 2 | 436 | 83 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5575683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655759.2 | XP_001655809.1 | 1 - 2 | 437 | 83 | |
XM_001655760.2 | XP_001655810.1 | 1 - 2 | 436 | 83 |