Assay Details
Gene Symbol: LOC5565878 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650196.2 | XP_001650246.1 | 2 - 3 | 296 | 103 | |
GenBank mRNA | AY433012.1 | - | 2 - 3 | 262 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
