Assay Details
Gene Symbol: LOC23687501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494709.2 | XP_011493011.1 | 2 - 3 | 355 | 69 | |
GenBank mRNA | AY431382.1 | - | 2 - 3 | 259 | 69 | |
DQ440475.1 | - | 1 - 2 | 151 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
