Assay Details
Gene Symbol: LOC5576426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662584.2 | XP_001662634.2 | 2 - 3 | 506 | 57 | |
GenBank mRNA | AY431418.1 | - | 1 - 2 | 198 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
