Assay Details
Gene Symbol: LOC5576357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656045.2 | XP_001656095.2 | 2 - 3 | 296 | 75 | |
GenBank mRNA | AY431526.1 | - | 2 - 3 | 304 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
