Assay Details
Gene Symbol: LOC5577626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846556.1 | XP_021702248.1 | 3 - 4 | 439 | 76 | |
XM_021846557.1 | XP_021702249.1 | 4 - 5 | 605 | 76 | ||
XM_021846558.1 | XP_021702250.1 | 3 - 4 | 299 | 76 | ||
XM_021846559.1 | XP_021702251.1 | 3 - 4 | 386 | 76 | ||
GenBank mRNA | AY431550.1 | - | 2 - 3 | 286 | 76 | |
DQ440291.1 | - | 2 - 3 | 254 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
