Assay Details
Gene Symbol: LOC5577626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846556.1 | XP_021702248.1 | 6 - 7 | 80 | ||
XM_021846557.1 | XP_021702249.1 | 7 - 8 | 80 | |||
XM_021846558.1 | XP_021702250.1 | 6 - 7 | 80 | |||
XM_021846559.1 | XP_021702251.1 | 6 - 7 | 80 | |||
GenBank mRNA | AY431550.1 | - | 5 - 6 | 713 | 81 | |
DQ440291.1 | - | 5 - 6 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
