Assay Details
Gene Symbol: LOC5573805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845622.1 | XP_021701314.1 | 3 - 4 | 1204 | 96 | |
XM_021845624.1 | XP_021701316.1 | 3 - 4 | 1206 | 96 |
