Assay Details
Gene Symbol: LOC5573805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845622.1 | XP_021701314.1 | 2 - 3 | 554 | 73 | |
XM_021845624.1 | XP_021701316.1 | 2 - 3 | 556 | 73 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5573805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845622.1 | XP_021701314.1 | 2 - 3 | 554 | 73 | |
XM_021845624.1 | XP_021701316.1 | 2 - 3 | 556 | 73 |