Assay Details
Gene Symbol: LOC5576270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662506.3 | XP_001662556.2 | 1 | 326 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001662506.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
