Assay Details
Gene Symbol: LOC5573814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661011.3 | XP_001661061.2 | 3 - 4 | 2801 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5573814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661011.3 | XP_001661061.2 | 3 - 4 | 2801 | 101 |
Set Membership: |
![]() ![]() ![]() ![]() |