Assay Details
Gene Symbol: LOC5570801 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653256.2 | XP_001653306.1 | 2 - 3 | 497 | 67 | |
XM_021849823.1 | XP_021705515.1 | 2 - 3 | 412 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
