Assay Details
Gene Symbol: LOC5570801 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653256.2 | XP_001653306.1 | 5 - 6 | 1175 | 93 | |
XM_021849823.1 | XP_021705515.1 | 5 - 6 | 1090 | 93 |
